accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2210-25-5

IUPAC Name: N-(propan-2-yl)prop-2-enamide
Synonyms: N-isopropylacrylamide
Molecular Weight (g/mol): 113.16
Molecular Formula: C6H11NO
InChi Key: QNILTEGFHQSKFF-UHFFFAOYSA-N
SMILES: CC(C)NC(=O)C=C
Boiling Point 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Physical Form 
  • (4)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)