accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClOBr

CAS RN 22118-09-8

IUPAC Name: 2-bromoacetyl chloride
Synonyms: 2-bromoacetyl chloride
Molecular Weight (g/mol): 157.39
Molecular Formula: C2H2BrClO
InChi Key: SYZRZLUNWVNNNV-UHFFFAOYSA-N
SMILES: ClC(=O)CBr
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)