accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OHNFFF

CAS RN 22123-19-9

IUPAC Name: 6-methyl-4-(trifluoromethyl)-1,2-dihydropyridin-2-one
Synonyms: 6-methyl-4-(trifluoromethyl)-1H-pyridin-2-one
Molecular Weight (g/mol): 177.13
Molecular Formula: C7H6F3NO
InChi Key: CDRMKCNOGIBXOG-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(=O)N1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results