accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2217-15-4

IUPAC Name: 1,4-bis(propan-2-yl) 2,3-dihydroxybutanedioate
Synonyms: diisopropyl tartrate
Molecular Weight (g/mol): 234.25
Molecular Formula: C10H18O6
InChi Key: XEBCWEDRGPSHQH-UHFFFAOYNA-N
SMILES: CC(C)OC(=O)C(O)C(O)C(=O)OC(C)C
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Boiling Point 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)