accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClNH3

CAS RN 2217-40-5

IUPAC Name: 1,2,3,4-tetrahydronaphthalen-1-aminium chloride
Synonyms: 1,2,3,4-tetrahydronaphthalen-1-aminium chloride
Molecular Weight (g/mol): 183.68
Molecular Formula: C10H14ClN
InChi Key: DETWFIUAXSWCIK-UHFFFAOYNA-N
SMILES: [Cl-].[NH3+]C1CCCC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (4)
  • (3)
Quantity 
  • (1)
  • (3)
  • (3)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (4)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)