accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 22179-86-8

H2NNOHFFF(Z)

CAS RN 22179-86-8

IUPAC Name: (Z)-N'-hydroxy-4-(trifluoromethyl)benzene-1-carboximidamide
Synonyms: (Z)-N'-hydroxy-4-(trifluoromethyl)benzenecarboximidamide
Molecular Weight (g/mol): 204.15
Molecular Formula: C8H7F3N2O
InChi Key: QCVFLUSIBKAKPC-UHFFFAOYSA-N
SMILES: N\C(=N/O)C1=CC=C(C=C1)C(F)(F)F
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results