accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2OFFFFFF

CAS RN 22227-26-5

IUPAC Name: 3,5-bis(trifluoromethyl)benzamide
Synonyms: 3,5-bis(trifluoromethyl)benzamide
Molecular Weight (g/mol): 257.14
Molecular Formula: C9H5F6NO
InChi Key: YNOPIKHMZIOWHS-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results