accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CdH3COOH3COO

CAS RN 2223-93-0

IUPAC Name: cadmium(2+) dioctadecanoate
Synonyms: cadmium stearate
Molecular Weight (g/mol): 679.37
Molecular Formula: C36H70CdO4
InChi Key: GWOWVOYJLHSRJJ-UHFFFAOYSA-L
SMILES: [Cd++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results