accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClCH3H2NBOHHO

CAS RN 22237-12-3

IUPAC Name: hydrogen (3-amino-4-methylphenyl)boronic acid chloride
Synonyms: hydrogen 3-amino-4-methylphenylboronic acid chloride
Molecular Weight (g/mol): 187.43
Molecular Formula: C7H11BClNO2
InChi Key: BKFDOPYYSBQWIK-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC1=CC=C(C=C1N)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results