CAS RN 22248-79-9
IUPAC Name:
(1E)-2-chloro-1-(2,4,5-trichlorophenyl)ethenyl dimethyl phosphate
Synonyms:
tetrachlorvinphos
Molecular Weight (g/mol):
365.95
Molecular Formula:
C10H9Cl4O4P
InChi Key:
UBCKGWBNUIFUST-BJMVGYQFSA-N
SMILES:
COP(=O)(OC)O\C(=C\Cl)C1=CC(Cl)=C(Cl)C=C1Cl
1
–
1
of
1
results
1
(Z)-Tetrachlorvinphos, TRC
CAS: 22248-79-9 Molecular Formula: C10 H9 Cl4 O4 P Molecular Weight (g/mol): 363.8992561 Synonym: Phosphoric acid, (1Z)-2-chloro-1-(2,4,5-trichlorophenyl)ethenyl dimethyl ester,Phosphoric acid, 2-chloro-1-(2,4,5-trichlorophenyl)ethenyl dimethyl ester, (Z)-,Phosphoric acid, 2-chloro-1-(2,4,5-trichlorophenyl)vinyl dimethyl ester, (Z)- (8CI),(Z)-Tetrachlorovinphos,CVMP,Debantic,Gardicide,Gardona,Rabon,Rabond,SD 8447,Stirofos,Stirophos,Stitifos,Tetrachlorvinfos,Tetrachlorvinphos,Vinfos,cis-Gardona IUPAC Name: [(Z)-2-chloro-1-(2,4,5-trichlorophenyl)ethenyl] dimethyl phosphate SMILES: COP(=O)(OC)O\C(=C/Cl)\c1cc(Cl)c(Cl)cc1Cl