accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 22268-65-1

IUPAC Name: 3-ethyl-2-[5-(3-ethyl-2,3-dihydro-1,3-benzothiazol-2-ylidene)penta-1,3-dien-1-yl]-1,3-benzothiazol-3-ium perchlorate
Synonyms: dithiazanine perchlorate
Molecular Weight (g/mol): 491.02
Molecular Formula: C23H23ClN2O4S2
InChi Key: HSVLFBSXAKAPBG-UHFFFAOYSA-M
SMILES: [O-][Cl](=O)(=O)=O.CCN1C(SC2=CC=CC=C12)=CC=CC=CC1=[N+](CC)C2=CC=CC=C2S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results