accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2227-79-4

IUPAC Name: benzenecarbothioamide
Synonyms: thiobenzamide
Molecular Weight (g/mol): 137.2
Molecular Formula: C7H7NS
InChi Key: QIOZLISABUUKJY-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results