accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClNNHNBr

CAS RN 22276-95-5

IUPAC Name: 5-bromo-4-chloro-7H-pyrrolo[2,3-d]pyrimidine
Synonyms: 5-bromo-4-chloro-7H-pyrrolo[2,3-d]pyrimidine
Molecular Weight (g/mol): 232.47
Molecular Formula: C6H3BrClN3
InChi Key: OXLMTRZWMHIZBY-UHFFFAOYSA-N
SMILES: ClC1=C2C(Br)=CNC2=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results