accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NI

CAS RN 22282-58-2

IUPAC Name: 2-iodo-3-methylpyridine
Synonyms: 2-iodo-3-methylpyridine
Molecular Weight (g/mol): 219.03
Molecular Formula: C6H6IN
InChi Key: PTBOOHAPESUXGV-UHFFFAOYSA-N
SMILES: CC1=CC=CN=C1I
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results