accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 22300-56-7

CH3NNNOO

CAS RN 22300-56-7

IUPAC Name: 5-methyl-2-phenyl-2H-1,2,3-triazole-4-carboxylate
Synonyms: 5-methyl-2-phenyl-1,2,3-triazole-4-carboxylate
Molecular Weight (g/mol): 202.19
Molecular Formula: C10H8N3O2
InChi Key: GQYQHAGPALBYDO-UHFFFAOYSA-M
SMILES: CC1=NN(N=C1C([O-])=O)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results