accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 22317-89-1

H3CSOOSOOCl

CAS RN 22317-89-1

IUPAC Name: methanesulfonylmethanesulfonyl chloride
Synonyms: methanesulfonylmethanesulfonyl chloride
Molecular Weight (g/mol): 192.63
Molecular Formula: C2H5ClO4S2
InChi Key: DQDYGBSUPOHHMW-UHFFFAOYSA-N
SMILES: CS(=O)(=O)CS(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results