accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 22325-27-5

CH3NSHNCH3

CAS RN 22325-27-5

IUPAC Name: 4,6-dimethyl-1,2-dihydropyrimidine-2-thione
Synonyms: 4,6-dimethyl-1H-pyrimidine-2-thione
Molecular Weight (g/mol): 140.2
Molecular Formula: C6H8N2S
InChi Key: RAFAYWADRVMWFA-UHFFFAOYSA-N
SMILES: CC1=CC(C)=NC(=S)N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results