accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 223437-11-4

IUPAC Name: 1-butyl-1-methylpyrrolidin-1-ium; trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Synonyms: 1-butyl-1-methylpyrrolidin-1-ium; bistriflylimide anion
Molecular Weight (g/mol): 422.4
Molecular Formula: C11H20F6N2O4S2
InChi Key: HSLXOARVFIWOQF-UHFFFAOYSA-N
SMILES: CCCC[N+]1(C)CCCC1.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results