accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 223499-20-5

IUPAC Name: 5-[1-methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl]thiophene-2-carboxylic acid
Synonyms: 5-[1-methyl-5-(trifluoromethyl)pyrazol-3-yl]thiophene-2-carboxylic acid
Molecular Weight (g/mol): 276.23
Molecular Formula: C10H7F3N2O2S
InChi Key: MGYVPTDPOYVOQK-UHFFFAOYSA-N
SMILES: CN1N=C(C=C1C(F)(F)F)C1=CC=C(S1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results