accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2235-00-9

IUPAC Name: 1-ethenylazepan-2-one
Synonyms: polyvinylcaprolactam
Molecular Weight (g/mol): 139.2
Molecular Formula: C8H13NO
InChi Key: JWYVGKFDLWWQJX-UHFFFAOYSA-N
SMILES: C=CN1CCCCCC1=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results