accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

O

CAS RN 2235-15-6

IUPAC Name: 1,2-dihydroacenaphthylen-1-one
Synonyms: 1-acenaphthenone
Molecular Weight (g/mol): 168.2
Molecular Formula: C12H8O
InChi Key: JBXIOAKUBCTDES-UHFFFAOYSA-N
SMILES: O=C1CC2=C3C1=CC=CC3=CC=C2
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results