accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2237-30-1

NH2N

CAS RN 2237-30-1

IUPAC Name: 3-aminobenzonitrile
Synonyms: M-aminobenzonitrile
Molecular Weight (g/mol): 118.14
Molecular Formula: C7H6N2
InChi Key: NJXPYZHXZZCTNI-UHFFFAOYSA-N
SMILES: NC1=CC=CC(=C1)C#N
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)