accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 22374-89-6

HClH3CNH2

CAS RN 22374-89-6

IUPAC Name: hydrogen 4-phenylbutan-2-amine chloride
Synonyms: hydrogen α-benzylisopropylamine chloride
Molecular Weight (g/mol): 185.7
Molecular Formula: C10H16ClN
InChi Key: LVQSUJANKVLTKT-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CC(N)CCC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results