CAS RN 22385-77-9
IUPAC Name:
1-bromo-3,5-di-tert-butylbenzene
Synonyms:
1-bromo-3,5-di-tert-butylbenzene
Molecular Weight (g/mol):
269.23
Molecular Formula:
C14H21Br
InChi Key:
BUOWTUULDKULFI-UHFFFAOYSA-N
SMILES:
CC(C)(C)C1=CC(=CC(Br)=C1)C(C)(C)C
1
–
1
of
1
results
1
1-Bromo-3,5-di-tert-butylbenzene, 99%
CAS: 22385-77-9 Molecular Formula: C14H21Br Molecular Weight (g/mol): 269.226 MDL Number: MFCD00796945 InChI Key: BUOWTUULDKULFI-UHFFFAOYSA-N Synonym: 1-bromo-3,5-di-tert-butylbenzene,3,5-di-tert-butylbromobenzene,3,5-di-t-butylbromobenzene,1-bromo-3,5-di-t-butylbenzene,1-bromo-3,5-ditert-butyl benzene,benzene, 1-bromo-3,5-bis 1,1-dimethylethyl,1,5-bis tert-butyl-3-bromobenzene,acmc-1cpbo,g00018-watson-int,3,5-di-t-butyl-bromobenzene PubChem CID: 620136 IUPAC Name: 1-bromo-3,5-ditert-butylbenzene SMILES: CC(C)(C)C1=CC(=CC(=C1)Br)C(C)(C)C