accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOCH3OOCH3

CAS RN 22398-14-7

IUPAC Name: 1,3-dimethyl 2-(methoxymethylidene)propanedioate
Synonyms: 1,3-dimethyl 2-(methoxymethylidene)propanedioate
Molecular Weight (g/mol): 174.15
Molecular Formula: C7H10O5
InChi Key: RHFZTBSULNJWEI-UHFFFAOYSA-N
SMILES: COC=C(C(=O)OC)C(=O)OC
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)