CAS RN 224323-50-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(±)-Linezolid, TRC
CAS: 224323-50-6 Molecular Formula: C16 H20 F N3 O4 Molecular Weight (g/mol): 337.35 Synonym: Racemic Linezolid,N-[[3-[3-Fluoro-4-(4-morpholinyl)phenyl]-2-oxo-5-oxazolidinyl]methyl]acetamide IUPAC Name: N-[[3-(3-fluoro-4-morpholino-phenyl)-2-oxo-oxazolidin-5-yl]methyl]acetamide SMILES: CC(=O)NCC1CN(C(=O)O1)c2ccc(N3CCOCC3)c(F)c2