accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COON

CAS RN 22446-12-4

IUPAC Name: 2-(4-methoxyphenoxy)acetonitrile
Synonyms: 2-(4-methoxyphenoxy)acetonitrile
Molecular Weight (g/mol): 163.18
Molecular Formula: C9H9NO2
InChi Key: SNADCCRKYFVIEJ-UHFFFAOYSA-N
SMILES: COC1=CC=C(OCC#N)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results