CAS RN 2245-38-7
IUPAC Name:
1,6,7-trimethylnaphthalene
Synonyms:
2,3,5-trimethylnaphthalene
Molecular Weight (g/mol):
170.26
Molecular Formula:
C13H14
InChi Key:
JBXULKRNHAQMAS-UHFFFAOYSA-N
SMILES:
CC1=CC=CC2=CC(C)=C(C)C=C12
1
–
3
of
3
results
1
1,6,7-Trimethylnaphthalene, TRC
CAS: 2245-38-7 Molecular Formula: C13 H14 Molecular Weight (g/mol): 170.25 Synonym: Naphthalene, 1,6,7-trimethyl- (6CI, 7CI, 8CI, 9CI, ACI),1,6,7-Trimethylnaphthalene (ACI),2,3,5-Trimethylnaphthalene,NSC 89511 IUPAC Name: 1,6,7-trimethylnaphthalene SMILES: Cc1cc2cccc(C)c2cc1C