accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2251-79-8

FFFNNHNN

CAS RN 2251-79-8

IUPAC Name: 5-[4-(trifluoromethyl)phenyl]-2H-1,2,3,4-tetrazole
Synonyms: 5-[4-(trifluoromethyl)phenyl]-2H-1,2,3,4-tetrazole
Molecular Weight (g/mol): 214.15
Molecular Formula: C8H5F3N4
InChi Key: CCVCHQBLMDMSNN-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(C=C1)C1=NNN=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results