accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2252-44-0

FFFOBr

CAS RN 2252-44-0

IUPAC Name: 1-bromo-3-(trifluoromethoxy)benzene
Synonyms: 1-bromo-3-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 241.01
Molecular Formula: C7H4BrF3O
InChi Key: WVUDHWBCPSXAFN-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=CC(Br)=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results