accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 22545-15-9

IUPAC Name: 2-(4-ethoxyphenyl)ethan-1-ol
Synonyms: 2-(4-ethoxyphenyl)ethanol
Molecular Weight (g/mol): 166.22
Molecular Formula: C10H14O2
InChi Key: CNMVSNTVPZWQMI-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(CCO)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results