CAS RN 22556-62-3
IUPAC Name:
sodium 3-(hydrogen phosphonatooxy)-2-hydroxypropyl octadec-9-enoate
Synonyms:
sodium 3-(hydrogen phosphonatooxy)-2-hydroxypropyl octadec-9-enoate
Molecular Weight (g/mol):
458.51
Molecular Formula:
C21H40NaO7P
InChi Key:
XGRLSUFHELJJAB-UHFFFAOYNA-M
SMILES:
[Na+].CCCCCCCCC=CCCCCCCCC(=O)OCC(O)COP(O)([O-])=O