accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3FFFFFFHO

CAS RN 225920-05-8

IUPAC Name: 1-[3,5-bis(trifluoromethyl)phenyl]ethan-1-ol
Synonyms: 1-[3,5-bis(trifluoromethyl)phenyl]ethanol
Molecular Weight (g/mol): 258.16
Molecular Formula: C10H8F6O
InChi Key: MMSCIQKQJVBPIR-UHFFFAOYNA-N
SMILES: CC(O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)