accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOONOO

CAS RN 22621-41-6

IUPAC Name: 2-(methoxycarbonyl)-6-nitrobenzen-1-olate
Synonyms: 2-(methoxycarbonyl)-6-nitrobenzenolate
Molecular Weight (g/mol): 196.14
Molecular Formula: C8H6NO5
InChi Key: NIBVYEHAFBEVFI-UHFFFAOYSA-M
SMILES: COC(=O)C1=CC=CC(=C1[O-])[N+]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)