accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2267-23-4

IUPAC Name: 2-nitro-4-(trifluoromethoxy)aniline
Synonyms: 2-nitro-4-(trifluoromethoxy)aniline
Molecular Weight (g/mol): 222.12
Molecular Formula: C7H5F3N2O3
InChi Key: YCGFVAPIBALHRT-UHFFFAOYSA-N
SMILES: NC1=CC=C(OC(F)(F)F)C=C1[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results