accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OH(R)NHO

CAS RN 22677-21-0

IUPAC Name: (4R)-4-hydroxypyrrolidin-2-one
Synonyms: (4R)-4-hydroxypyrrolidin-2-one
Molecular Weight (g/mol): 101.11
Molecular Formula: C4H7NO2
InChi Key: IOGISYQVOGVIEU-GSVOUGTGSA-N
SMILES: O[C@H]1CNC(=O)C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results