accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 226878-01-9

NHONN

CAS RN 226878-01-9

IUPAC Name: N-(adamantan-1-yl)quinoxaline-2-carboxamide
Synonyms: N-(adamantan-1-yl)quinoxaline-2-carboxamide
Molecular Weight (g/mol): 307.4
Molecular Formula: C19H21N3O
InChi Key: ZKFVOZCCAXQXBU-UHFFFAOYSA-N
SMILES: O=C(NC12CC3CC(CC(C3)C1)C2)C1=CN=C2C=CC=CC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results