accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2270-59-9

CH3CH3Br

CAS RN 2270-59-9

IUPAC Name: 5-bromo-2-methylpent-2-ene
Synonyms: 5-bromo-2-methylpent-2-ene
Molecular Weight (g/mol): 163.06
Molecular Formula: C6H11Br
InChi Key: UNXURIHDFUQNOC-UHFFFAOYSA-N
SMILES: CC(C)=CCCBr
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results