accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 22720-75-8

H3COS

CAS RN 22720-75-8

IUPAC Name: 1-(1-benzothiophen-2-yl)ethan-1-one
Synonyms: 1-(1-benzothiophen-2-yl)ethanone
Molecular Weight (g/mol): 176.23
Molecular Formula: C10H8OS
InChi Key: SGSGCQGCVKWRNM-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC2=CC=CC=C2S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results