accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SnCH3O

CAS RN 2273-45-2

IUPAC Name: dimethylstannanone
Synonyms: stannane, dimethyloxo-
Molecular Weight (g/mol): 164.78
Molecular Formula: C2H6OSn
InChi Key: WNVQCJNZEDLILP-UHFFFAOYSA-N
SMILES: C[Sn](C)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results