accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClSnHH3CCH3H3C

CAS RN 2279-76-7

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 285.44
Molecular Formula: C9H23ClSn
InChi Key: ZXVSRBCDRCJMBB-UHFFFAOYSA-N
SMILES: Cl.CCC[SnH](CCC)CCC
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (6)
  • (6)