accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 22791-64-6

IUPAC Name: 2,4-dihydro-1,3-benzodioxin-6-amine
Synonyms: 2,4-dihydro-1,3-benzodioxin-6-amine
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: MUSIFRYVOVSNMY-UHFFFAOYSA-N
SMILES: NC1=CC=C2OCOCC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results