accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOClNSH3CCl

CAS RN 227958-96-5

IUPAC Name: ethyl 4,6-dichloro-2-(methylsulfanyl)quinoline-3-carboxylate
Synonyms: ethyl 4,6-dichloro-2-(methylsulfanyl)quinoline-3-carboxylate
Molecular Weight (g/mol): 316.2
Molecular Formula: C13H11Cl2NO2S
InChi Key: NRGNGBKXHHRCBD-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(SC)N=C2C=CC(Cl)=CC2=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results