accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClOSHN

CAS RN 22876-19-3

IUPAC Name: 5-chloro-2,3-dihydro-1,3-benzoxazole-2-thione
Synonyms: 5-chloro-3H-1,3-benzoxazole-2-thione
Molecular Weight (g/mol): 185.63
Molecular Formula: C7H4ClNOS
InChi Key: BOBIZYYFYLLRAH-UHFFFAOYSA-N
SMILES: ClC1=CC=C2OC(=S)NC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results