accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2ClNCl

CAS RN 22889-78-7

IUPAC Name: 3,5-dichloropyridin-4-amine
Synonyms: 3,5-dichloropyridin-4-amine
Molecular Weight (g/mol): 163
Molecular Formula: C5H4Cl2N2
InChi Key: ISIQAMHROGZHOV-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C=NC=C1Cl
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)