CAS RN 229-87-8
IUPAC Name:
phenanthridine
Synonyms:
phenanthridine 3,4-benzoquinoline 3,4-benzoisoquinoline 6-phenanthridine benzo(c)quinoline
Molecular Weight (g/mol):
179.22
Molecular Formula:
C13H9N
InChi Key:
RDOWQLZANAYVLL-UHFFFAOYSA-N
SMILES:
C1=CC=C2C(=C1)C=NC1=CC=CC=C21
1
–
1
of
1
results
1
Promotions Available
Phenanthridine, 98%
Phenanthridine, 98%
CAS: 229-87-8 Molecular Formula: C13H9N Molecular Weight (g/mol): 179.22 MDL Number: MFCD00004989 InChI Key: RDOWQLZANAYVLL-UHFFFAOYSA-N Synonym: 6-phenanthridine,benzo c quinoline,3,4-benzoquinoline,9-azaphenanthrene,3,4-benzoisoquinoline,5-azaphenanthrene,unii-62qgs7cps6,ccris 1234,62qgs7cps6 PubChem CID: 9189 ChEBI: CHEBI:36421 IUPAC Name: phenanthridine SMILES: C1=CC=C2C(=C1)C=NC3=CC=CC=C23