accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 22913-26-4

CH3OOS

CAS RN 22913-26-4

IUPAC Name: methyl thiophene-3-carboxylate
Synonyms: methyl thiophen-3-carboxylate
Molecular Weight (g/mol): 142.17
Molecular Formula: C6H6O2S
InChi Key: ZTRAEMILTFNZSM-UHFFFAOYSA-N
SMILES: COC(=O)C1=CSC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results