accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 22929-52-8

IUPAC Name: oxolan-3-one
Synonyms: dihydrofuran-3-one
Molecular Weight (g/mol): 86.09
Molecular Formula: C4H6O2
InChi Key: JLPJFSCQKHRSQR-UHFFFAOYSA-N
SMILES: O=C1CCOC1
Quantity 
  • (3)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results