accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 22931-71-1

H3COONOCH3Cl

CAS RN 22931-71-1

IUPAC Name: ethyl 4-chloro-6-methoxyquinoline-3-carboxylate
Synonyms: ethyl 4-chloro-6-methoxyquinoline-3-carboxylate
Molecular Weight (g/mol): 265.69
Molecular Formula: C13H12ClNO3
InChi Key: QURGQUFEJWWDRF-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN=C2C=CC(OC)=CC2=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results