accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SHN

CAS RN 2295-35-4

IUPAC Name: pyrrolidine-2-thione
Synonyms: 2-azacyclopentanthione
Molecular Weight (g/mol): 101.17
Molecular Formula: C4H7NS
InChi Key: IMWUREPEYPRYOR-UHFFFAOYSA-N
SMILES: S=C1CCCN1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)